Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Talon® Frame Connector
Supplier: VWR
VWR® Talon®
80862638
60097-055CS 2415.85 USD InStock
60097-055
VWR® Talon® Frame Connector
Support Rods and Frames
Frame connector with improved contours delivers simplicity, superior strength and a secure grip on lattice frame rods without slippage.
  • Compact design maximizes lab frame space
  • Independent rod adjustment
  • Simple installation of lattices
  • Smooth, rounded surfaces
  • Easy to clean
Angled adjustment screws allow easy setup and prevent misalignment. Two U-shaped slots and center through hole allow rods to be joined in 90° corner joint or 90° cross-connection configurations. Each rod adjusts independently for easy installation of lattices. Cast aluminum construction for superior strength. Silicone finish for maximum protection against corrosion. Smooth, rounded surfaces are easy to clean. Grips rods up to 13 mm (1/2") in diameter.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty