Would you like to log in again?
Support Rod Holder, Humboldt
Supplier: Humboldt
H-21160
60118-009EA 84.53 USD InStock
60118-009
Support Rod Holder, Humboldt
Support Stands
Consists of a heavy, cast-iron clamp protectively enameled, for fastening to edge of table, bench or shelf up to 38mm (11/2") thick
These support rod holder are made of steel.
Accommodates two 91/2mm (3/8") diameter support rods at right angles. With one 91/2mm (3/8") diameter by 508mm (20") plated steel rod.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty