Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Talon® Shelf-Grip Support

Supplier: VWR
VWR® Talon®
80863383
60119-001EA 339.36 USD InStock
60119-001
VWR® Talon® Shelf-Grip Support
Support Rods and Frames
Aluminum bracket with 140 mm (51/2") arm fastens quickly and firmly to any convenient shelf.

  • Aluminum construction
  • Supplied without rod
  • Depth of grip: 50 mm (2")

Accepts 13 mm (1/2") rods vertically and horizontally, to which ordinary rings and clamps attach, leaving bench surface clear of apparatus.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty