Would you like to log in again?
VWR® Talon® Gas Cylinder Stand
Supplier: VWR
VWR® Talon®
30400023
60141-000EA 790 USD InStock
60141-000
VWR® Talon® Gas Cylinder Stand
Support Stands
Permits convenient, safe cylinder transfer and upright support.
  • Heavy-duty cast iron construction
  • Rubber support wheels
  • Fully mobile
  • Secure fit around a range of tank diameters
Designed in two halves that fit securely around any tank from 17.8 – 23.5 cm (7 – 91/4") in diameter.
Made of heavy-duty cast iron, support has rubber wheels that convert it into a dolly for complete mobility. Can be tipped to approximately 35° and returned to vertical position.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty