Would you like to log in again?
EasyLok™ HPLC Fittings, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
EasyLok™
206160
60362-334PK 82.68 USD InStock
60362-334
EasyLok™ HPLC Fittings, Trajan Scientific and Medical
Fittings and Connectors
EasyLok™ fittings are composed of a knurled stainless steel nut and a double ended PEEK™ ferrule
  • Double ended ferrule seals at two points
  • Knurled to make finger tightening easy
These fittings are compatible with any standard female HPLC fitting including Swagelok®, Parker™, Waters®, Valco®, and Whatman®.
The PEEK™ ferrule slides over any 1/16" OD tubing to its required position, while the nut is finger tightened. Unlike stainless steel, the PEEK ferrule will not crush the tubing and can be easily readjusted for quick column changes. The unique double ended ferrule design seals at two points to prevent leaks. The fittings are compatible with any standard female HPLC fitting, including Swagelok®, Parker™, Waters®, Valco® and Whatman®. EasyLOK fittings are recommended for use with SGE GLT™ columns.
Certifications: RoHS compliant per Directive 2011/65/EU.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty