Would you like to log in again?
Microliter™ 7000 Series Syringes, Hamilton
Supplier: Hamilton
MICROLITER®
86200 88411 88500 88400 88011 86211 88000 88511 86250 80135 86257 80100 86259
60374-012EA 176.25 USD InStock
60374-012 60374-010 60374-006 60374-004 60374-008 60375-045 60375-078 60375-067 60375-089 60375-012 60375-023 60375-034 60375-158
Microliter™ 7000 Series Syringes, Hamilton
Syringes Syringes for Chromatography
7000 Series syringes feature a unique modified Microliter™ design based on the dead volume-free principle of positive displacement
Syringe Termination: A knurled hub is used exclusively on 7000 Series syringes. The hub handles up to 6000psig maximum injection pressure. The needle is removable, but with a limited number of gauges available because the plunger is fitted inside the needle. Autoclavable when disassembled. Gas sterilizable. Repeated autoclaving will shorten syringe life.
Needle Point Styles:
Point style 2 is a sharp, beveled, curved, non-coring needle point recommended for septum penetration.
Point style 3 is a blunt needle point for use with HPLC injection valves and for sample pipetting.
A fine tungsten wire plunger extends through the whole length of the syringe through to the needle end. Ideal for dispensing volumes from 0.05μL up to 5μL. Fluid path includes SST and tungsten.
Ordering information: For available needle gauges, lengths, and point styles to optimize the syringe for custom applications, contact your Avantor representative.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty