Would you like to log in again?
GC Septa, Hamilton Company
Supplier: Hamilton
76006 75801 76016 76010 75803 75806 76003 75813 76002 76019 76015 76017 75805 75804 75802 76020 76007 76004 76005 76009 76013 76022 76001 75810 75807
89220-878PK 29.04 USD InStock
89220-878 89220-876 60379-909 89220-874 89220-872 89220-894 89220-892 89220-870 89220-890 60379-749 60379-884 60379-862 60379-545 60379-589 60379-501 60379-964 60379-603 60379-625 60379-669 89220-888 89220-886 60379-760 89220-884 89220-882 89220-880
GC Septa, Hamilton Company
Septa
Hamilton Company offers the most comprehensive selection of products in the industry.
  • Offers a variety of septa for many GC
  • Each is manufactured to achieve the highest level of quality possible
High temperature septa is made of preconditioned PTFE/silicone rubber and is 3.175 mm (0.125") thick. The average life of these septa is up to 100 injections with a 26s gauge needle. The maximum operating temperature is 212 °C.
Three Layer Septa is made of preconditioned silicone rubber and is 3.81 mm (0.150") thick. The average life of these septa is up to 100 injections with a 26s gauge needle. The maximum operating temperature is 200 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty