Would you like to log in again?
VWR® Culture Tubes, Plastic, with Dual-Position Caps
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
3328-355-300 3315-800-000 3335-335-300 3329-355-300 3316-800-000 3529-355-300 3536-335-300 3515-800-000 3336-335-300 3528-355-300 3535-335-300 3516-800-000
60818-565CS 244.86 USD InStock
60818-565 60818-576 60818-664 60818-500 60818-689 60818-667 60818-496 60818-661 60818-419 20170-579 60818-725 60818-703
VWR® Culture Tubes, Plastic, with Dual-Position Caps
Tubes Culture Tubes
Choose from clear polystyrene or natural polypropylene disposable tubes.
  • Available in polystyrene or polypropylene formats
  • Supplied with dual-position caps to facilitate sample aspiration
Polypropylene tubes are autoclavable to 122 °C (252 °F) and freezable to −90 °C (−194 °F). Tubes feature dual-position polyethylene caps (not autoclavable). Dual-position caps permit sample aspiration.
Ordering information: Caps for nonsterile tubes are packed separately.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty