Would you like to log in again?
Polystyrene Test Tube, 13×75, Globe Scientific
Supplier: Globe Scientific
110470
60819-970PK 59.5 USD InStock
60819-970
Polystyrene Test Tube, 13×75, Globe Scientific
Tubes Test Tubes
Polystyrene tubes can withstand 1500 rpm during centrifugation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty