Would you like to log in again?
Transport Tube, 50 ml, Globe Scientific
Supplier: Globe Scientific
6255 6256
60819-978CS 339.49 USD InStock
60819-978 60819-976
Transport Tube, 50 ml, Globe Scientific
Tubes Test Tubes
These 50 ml screw cap tubes are leak-resistant and ideal for centrifugation, transport, storage, and specimen collection
Tubes have a conical bottom with a skirt and feature white printed graduation marks and a white frosted writing area.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty