Would you like to log in again?
Flat Bottom Test Tubes, Globe Scientific
Supplier: Globe Scientific
115060 114010
60819-988CS 118.53 USD InStock
60819-988 10014-270
Flat Bottom Test Tubes, Globe Scientific
Tubes Test Tubes
Ideal for use as a secondary tube for barcoding on automated analyzers.
  • Flat bottom, self-standing tubes
  • Available in clear polystyrene or unbreakable polypropylene
The 3 ml polystyrene tubes are graduated with a rim.
Popular brands of analyzers include Hitachi®, Olympus®, Corning®, Ortho®, Kodak®, Beckman®, IA®, Tosoh®, Vitros® and more.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty