Would you like to log in again?
Tiered Plug Stopper Closures, Polyethylene, Globe Scientific
Supplier: Globe Scientific
118127Y 118127G 118127C 118127B 118127R
60820-144PK 32.32 USD InStock
60820-144 60820-146 60820-148 60820-140 60820-142
Tiered Plug Stopper Closures, Polyethylene, Globe Scientific
Closures Test/Sample Tube Closures
Polyethylene flanged plug caps provide exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes
Closures fit 12mm tubes, and are available in a variety of colors to simplify identification and selection.
They feature two flexible flanges that press firmly against the inside of the tube wall, thereby providing better leak-resistance than standard plug caps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty