Would you like to log in again?
Corning® Cap, Phenolic, PTFE Liner
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
9998-15 9998-18 9998-13 9998-241 9998-24
66110-220CS 499.76 USD InStock
66110-220 60827-225 60827-229 60827-228 60827-227
Corning® Cap, Phenolic, PTFE Liner
Closures Test/Sample Tube Closures
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving.
  • Available in different sizes
Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty