Would you like to log in again?
Corning® Reusable Phenolic Caps, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
9999-182 9999-38 9999-281 9999-152 9999-24 9999-241 9999-132 9999-381M
60827-250CS 549.25 USD InStock
60827-250 60827-294 60827-409 60827-271 60827-318 60827-330 60827-351 60827-237
Corning® Reusable Phenolic Caps, Corning
Closures Bottle Closures
These reusable Corning® caps are fabricated from specially formulated phenolic resin to resist the effects of temperature and steam in autoclaving.
Caps have GPI threads and are provided with a glued-in white rubber liner.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty