Would you like to log in again?
PYREX® Flat Bottom Culture Tubes, Rimless, Corning
Supplier: Corning
PYREX®
9850-25 9850-25X 9850-25XX
60856-132CS 282.41 USD InStock
60856-132 60856-130 60856-134
PYREX® Flat Bottom Culture Tubes, Rimless, Corning
Tubes Culture Tubes
Flat-bottom culture tubes are ideal for plant tissue and general culture work.
Two-position polypropylene closures are sold separately. Position closure open for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.
The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage.
Ordering information: Tubes and closures sold separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty