Would you like to log in again?
Corning® Polypropylene Closures, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
26797-38 26796-38 26799-40 26795-25 26798-40
60856-142CS 362.14 USD InStock
60856-142 60856-147 60856-145 60856-149 60856-140
Corning® Polypropylene Closures, Corning
Closures Test/Sample Tube Closures
These Corning® polypropylene closures are designed for culture tubes and flasks.
  • Internal drip ring minimizes contamination
  • Autoclavable at 121 °C
Closure offers two positions: open for gas exchange or closed for humidified environment.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty