Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Petri Dishes, Sterile, Simport
Supplier: Simport
D210-15 D210-8 D210-8 D210-7WL D210-7 D210-14 D210-13 D210-13
60872-306CS 300.24 USD InStock
60872-306 60872-308 CA25378-105 60872-314 60872-484 CA73960-176 76299-234 60872-482
Petri Dishes, Sterile, Simport
Culture Dishes Petri Dishes
These Petri dishes are available in a variety of shapes and sizes for use in routine procedures and with automated equipment.
  • Economical
  • Optically clear
  • Precision molded from biomedical grade polystyrene so cultures are clearly visible without distortion
Model 100×20 mm provides with raised straight ridge around top for stable stacking for unimpaired observation of specimen growth.
Model 100×20 mm with three venting ribs on the underside of the lid to prevent condensation build-up.
Model 100×25 mm accomodates deeper fills for longer culture periods. Use for fungal cultures and plant propagation. This dish has three venting ribs on the underside of the lid to prevent condensation build-up.
Model 60×15 mm is for use whenever a small quantity of culture is desirable. For unimpaired observation of specimen growth with raised straight ridge around top for stable stacking. This dish has three venting ribs on the edge of the dish to prevent condensation build-up.
Model 50×9 mm is a box-type dish for classroom studies, water studies, culturing of mycobacteria, aerosol testing, membrane filtering, and immunodiffusion techniques. Tight lid prevents sample dehydration.
Model 35×10 mm is ideal for small quantities of culture media. For unimpaired observation of specimen growth with raised straight ridge around top for stable stacking. This dish has three venting ribs on the underside of the lid to prevent condensation build-up.
Packaging: Dishes are packaged in heavy-wall polyethylene sleeves.
Caution: Dishes are not designed for use in tissue culture applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty