Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sample Tubes with External Threads, Simport Scientific

Supplier: Simport

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

T501-5TPR T501-4AT T501-1ATPR T501-4ATPR T501-2ATPR T501-3AT T501-2AT T501-3AT T501-4T T501-2T T501-2AT T501-4TPR T501-5AT T501-4AT T501-4ATPR T501-5T T501-5ATPR T501-5T T501-5TPR T501-2T T501-5ATPR T501-4TPR T501-1ATPR T501-2TPR T501-1AT T501-2ATPR T501-2TPR T501-4T T501-3ATPR T501-5AT
60872-440CS 328.79 USD InStock
60872-440 CA60872-350 89135-598 CA60872-352 60872-442 CA60872-348 60872-438 60872-434 CA60872-344 60872-436 CA60872-346 60872-430 CA60872-340 60872-452 CA60872-342 60872-450 CA89135-598 490010-468 CA60872-336 10154-264 60872-426 CA60872-338 60872-428 CA11006-402 CA60872-354 60872-444 CA60872-334 CA60872-433 76299-336 CA11027-092
Sample Tubes with External Threads, Simport Scientific
Tubes Test Tubes
Vial has external threads, providing a smooth and uniform inner surface, thus reducing the risk of contamination.

  • Manufactured from non-toxic polypropylene
  • Height of tube is with cap
  • Caps available with or without silicone washer
  • Caps feature a long skirt and super-fast thread design that allows for single turn seal or removal
  • Tube and cap are made of polypropylene, providing the same coefficient of expansion

Designed for the storage and transportation of biological material. The tube provides strength and clarity and exhibits some unique design features. Tubes can be autoclaved (121 °C) in upright position with caps loosened. Round bottom tubes can only be centrifuged up to 17000 ×g.

Certifications: 95 kPa tested.

Ordering information: Screw caps must be ordered separately.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty