Would you like to log in again?
Screw Caps for T501 Sample Tubes, Simport Scientific
Supplier: Simport
T502B T502NSP T501Y T501L T502W T501B T502N T502BSP T501R T502B T501GY T501W T501O T501GY T501R T502W T502-G T501O T502R T501N T502Y T502YSP T502Y T501DG T502N T502R T501N T502L T501Y T502DG T502L T501L T502O T501W T501B
60872-462CS 115.39 USD InStock
60872-462 CA60872-372 CA60872-374 60872-464 60872-460 76299-342 76003-510 76299-340 60872-456 CA60872-366 76003-810 76003-514 60872-458 CA60872-368 76299-346 76299-344 CA60872-362 60872-474 CA60872-384 CA60872-386 60872-476 60872-454 CA60872-364 CA60872-380 60872-470 60872-472 CA60872-382 CA60872-360 CA60872-358 CA11014-056 60872-466 CA60872-376 CA60872-378 CA60872-356 76299-338
Screw Caps for T501 Sample Tubes, Simport Scientific
Closures Test/Sample Tube Closures
Caps are available either with or without a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures.
  • Made of polypropylene
  • Available with or without silicone washer
  • Long skirt and super fast thread design for easy seal
  • Autoclavable
As for tubes, these closures are also made of polypropylene, providing the same coefficient of expansion for both, which further enhances the leakproof qualities of the vials at changing temperatures. The cap features a long skirt and a super-fast thread design that allows it to be removed or sealed with a single turn.
Ordering information: Tubes T501 series sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty