Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amplitube™ PCR Reaction Tubes, 0.5 ml, Simport Scientific

Supplier: Simport

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Amplitube™
T325-4N T325-3R T325-3B T325-4G T325-3N T325-4B T325-3B T325-3R T325-4B T325-3G T325-3Y T325-4Y T325-4Y T325-3Y T325-4G T325-4R T325-3N T325-3G T325-4R T325-4N
89429-028PK 78.51 USD InStock
CA48648-136 CA48648-134 CA48648-132 CA48648-131 CA48648-130 CA48648-140 89429-028 89429-026 89429-030 60872-624 60872-616 60872-626 89429-024 60872-620 89429-032 CA11028-434 60872-618 CA48648-139 CA48648-138 CA48648-137
Amplitube™ PCR Reaction Tubes, 0.5 ml, Simport Scientific
Tubes PCR Tubes
Tubes are made of transparent superior grade polypropylene for improved viewing of the sample contents.

  • Attached hinged caps are either dome or flat-topped
  • Caps provide positive sealing during thermal cycling and prevent evaporation
  • Caps can be easily opened and closed with one hand
  • Graduated in 0.1 ml increments
  • Autoclavable

Tubes also feature an integral shield that prevents contamination with surface of lid. A frosted writing surface is provided for sample identification.

Ultra-thin wall design ensures rapid thermal transfer and significant reduction in cycle and PCR reaction time.

Rnase-, Dnase-, Pyrogen-, and DNA-free.

Ordering information: Available in colorless or four non-cytotoxic colors.

Packaging: Packaged in tamper-proof, resealable safety-lock bags.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty