Would you like to log in again?
Amplitube™ PCR Reaction Tubes, 0.2 ml, Simport Scientific
Supplier: Simport
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Amplitube™
T325-2VN T325-2VR T325-1B T325-2N T325-2VY T325-2Y T325-2VB T325-2G T325-1VR T325-1G T325-2R P200-601S T325-2VB T325-2R T325-1N T325-1G T325-1VG T325-2VR T325-2VG T325-2G T325-2VG T325-1B T325-2N T325-1VY T325-1R T325-2VN T325-1VB T325-2B T325-2B T325-1VN T325-1N T325-1Y T325-2VY T325-1Y T325-1R T325-2Y
89429-016PK 63.86 USD InStock
CA10030-910 89429-016 89429-014 10030-940 89429-018 60872-636 89429-020 60872-638 60872-632 89429-022 CA10030-908 CA10030-906 CA48648-129 CA48648-128 CA48648-126 CA60872-628 CA48648-125 10030-936 10030-914 CA48648-124 CA48648-145 10030-916 10030-938 CA48648-144 10030-910 CA48648-143 CA48648-142 CA10030-940 10030-934 CA48648-141 10030-912 60872-630 10030-906 60872-628 CA10030-938 10030-908
Amplitube™ PCR Reaction Tubes, 0.2 ml, Simport Scientific
Tubes PCR Tubes
Amplitube™ PCR Reaction tubes are made of transparent superior grade polypropylene for improved viewing of the sample contents.
  • Attached hinged caps are either dome or flat-topped
  • Caps provide positive sealing during thermal cycling and prevent evaporation
  • Caps can be easily opened and closed with one hand
  • Autoclavable
Tubes are available with or without an integral shield that prevents contamination with surface of lid. A frosted writing surface is provided for sample identification.
Ultra-thin wall design ensures rapid thermal transfer and significant reduction in cycle and PCR reaction time.
Rnase-, Dnase-, Pyrogen-, and DNA-free.
Ordering information: Available in colorless or four non-cytotoxic colors.
Packaging: Packaged in tamper-proof, resealable safety-lock bags.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty