Would you like to log in again?
Nalgene® Unwire™ Racks, ResMer® Resin, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene® Unwire™
5970-0516 5970-0430 5970-0225 5970-0116 5970-0230 5970-0020 5970-0016 5970-0513 5970-0113 5970-0313 5970-0416 5970-0425 5970-0413 5970-0530 5970-0213 5970-0520 5970-0420 5970-0316 5970-0525 5970-0030 5970-0120 5970-0125 5970-0025 5970-0216 5970-0130 5970-0220 5970-0013 5970-0325 5970-0320 5970-0330
60914-718CS 298.43 USD InStock
60914-718 60914-616 60914-638 60914-875 60914-730 60914-977 60914-707 60914-729 60915-073 60914-649 60914-627 60915-051 60914-605 60915-095 60915-030 60914-809 60915-131 60915-110 60914-886 60914-820 60914-842 60914-864 60914-762 60914-740 60914-944 60914-988 60914-922 60914-900 60914-966 60914-660
Nalgene® Unwire™ Racks, ResMer® Resin, Thermo Scientific
Racks Tube Racks
These durable, brightly colored racks feature a three-tier design
Racks for 13mm and 16mm tubes hold 72 tubes; racks for 20mm and 25mm tubes hold 40 tubes; racks for 30mm tubes holds 24 tubes. Autoclavable.
They have excellent chemical resistance and can be immersed in a water bath. Letters and numbers on top tier aid in tube identification. Labels can be placed on large end plates. ResMer® technology allows these racks to be used over a wide temperature range.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty