Would you like to log in again?
SP Bel-Art No-Wire™ Test Tube Racks, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
No-Wire™
187460003 F18745-0003 187450002 F18746-0002 187460004 F18746-0001 187470000 F18745-0004 187470003 187470001 F18745-0001 187450000 F18747-0004 187460000
60915-410CS 266.6 USD InStock
60915-410 60915-411 60915-422 60915-400 60915-412 60915-425 60915-414 60915-404 60915-405 10153-666 60915-417 60915-406 60915-418 60915-408
SP Bel-Art No-Wire™ Test Tube Racks, Polypropylene, Bel-Art Products, a part of SP
Racks Tube Racks
A great alternative to wire racks, with price- and space-saving advantages over other plastic racks.
  • Blend of polypropylene construction
  • Autoclavable
  • Meets the demands of daily usage in the lab
  • Available in numerous hole sizes
  • Choice of three colors for safety and identification
  • Features end plates for labeling; numbered and lettered rows for tube identification
  • Interlocking feet allow stacking for storage
  • Steam autoclavable at 121 °C (250 °F)
Made of a special blend of autoclavable polypropylene; will sink in a water bath and maintain stability, even in agitated water.
Dimensions for all racks are identical, and they assemble in seconds.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty