Would you like to log in again?
SP Bel-Art Heavy Duty Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F18856-0000 F18858-0000 F18859-0000 F18855-0000 F18857-0000
60916-382CS 323.74 USD InStock
60916-382 60916-407 60916-429 60916-349 60916-305
SP Bel-Art Heavy Duty Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
These heavy-duty 3 tier racks are stackable and compact, and have hand grips in the end plates for easy transport.
  • Compact design
  • Polypropylene construction
  • Autoclavable
  • Stackable
The base tier has rounded wells with drain holes, and the top tier has lettered and numbered rows. Molded of polypropylene they are steam autoclavable at 121ºC (250ºF). All racks are 21.3 cm (8³/₈") long x 11 cm (4⁵/₁₆") wide.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty