Would you like to log in again?
VWR® Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
Supplier: VWR
VWR®
F384VWR100 F96VWR100
60941-126CS 109.15 USD InStock
60941-126 60941-124
VWR® Aluminium Foil Seals for PCR and Storage (96- and 384-Well Plates)
Seals Microplate Seals
Aluminium foils, 75.7 µm thick, for use with 96- or 384-well plates. Fit inside the rim of raised rim plates. These foils have one partial-width end tab. Available non sterile only.
  • Recommended for temperatures from −80 to +120 °C
  • Certified free from DNase, RNase and nucleic acids
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty