Would you like to log in again?
SP Bel-Art Poxygrid® Half-Size Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Poxygrid®
F18788-2000 F18788-1600 F18788-2001 F18788-1301 F18788-2000 F18788-2003 F18788-1603 F18788-1601 F18788-1300
60941-583CS 1518.52 USD InStock
60941-583 60941-572 60941-594 60941-561 60941-619 60941-608 60941-621 490012-480 60941-596
SP Bel-Art Poxygrid® Half-Size Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Excellent space-savers for use in water baths, laminar flow hoods, fume hoods or any place where space is limited.
  • Compact racks
  • Epoxy-coated steel wire construction
  • Chemical and corrosion-resistant
  • Unaffected by organic solvents, salts, weak acids, and weak alkalis
  • Autoclavable
These compact racks have square holes, three decks, slotted mesh bottom deck, and reinforced edges.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty