Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® Racks, Polycarbonate, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nalgene®
5929-0020 5929-0030 5929-0016
60985-148CS 430.65 USD InStock
60985-148 60985-159 60985-137
Nalgene® Racks, Polycarbonate, Thermo Scientific
Racks Tube Racks
Racks are ideal for use below 4°C (39.2°F)

Note: Autoclavable, but repeated autoclaving may reduce mechanical strength.

They are made of transparent, rustproof, and corrosion-resistant polycarbonate. Holes are keyed with numbers and letters for easy identification. Wide spacing allows convenient insertion and removal of tubes. Racks stack when empty for convenient storage. Will not float in water baths.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty