Would you like to log in again?
VWR® Tube Racks for Radioimmunoassay Tubes, PP
Supplier: VWR
VWR®
F18871-0000 F18870-0000 F18872-0000
60985-319CS 526.58 USD InStock
60985-319 60985-283 60985-250
VWR® Tube Racks for Radioimmunoassay Tubes, PP
Racks Tube Racks
These polypropylene (PP) plastic racks provide a practical method of pipetting and storing a large number of samples.
  • Organize over 100 radioimmunoassay samples in one rack
  • Useful in clinical labs for storage of samples
  • Steam autoclavable at 121°C (250°F)
By using alternate rows and rotating the tubes, it is easy to keep track of the last tube filled.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty