Would you like to log in again?
Nalgene® Peg Racks, Polypropylene, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
5977-0013 5977-0313 5977-0317 5977-0017
60985-406CS 162.01 USD InStock
60985-406 60985-421 60985-400 60985-415
Nalgene® Peg Racks, Polypropylene, Thermo Scientific
Racks Draining Racks
Racks hold sample collection tubes, reaction tubes, and test tubes
Racks consist of an array of pegs spaced to hold test tubes upright and display the contents. Pegs are also excellent for holding inverted tubes for draining/drying.
Also hold and dry electrophoresis and chromatography plates. Durable, chemical-resistant, and autoclavable. Racks sink in water bath.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty