Would you like to log in again?
SP Bel-Art Serum Vial Racks, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F18513-1640 F18513-2040 F18513-0072
60985-476EA 45.12 USD InStock
60985-476 60985-498 60985-454
SP Bel-Art Serum Vial Racks, Polypropylene, Bel-Art Products, a part of SP
Racks Chromatography Vial Racks
Just the right size for serum vials, RIA vials and T3/T4 reaction vials.
  • Polypropylene rack fabrication
  • Corrosion-resistant
  • Long effective lifepsan
  • Autoclavable
Long-lasting and corrosion-resistant, these polypropylene racks fit most water baths for elevated temperature incubations. Top shelf height is 41 mm (158"). End plates have handles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty