Would you like to log in again?
Yeast Morphology Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M138-500G M138-100G M138-500G M138-100G
61000-020EA 131.68 USD InStock
61000-020 95021-054 CA95021-054 CA61000-020
Yeast Morphology Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Yeast Morphology Agar is recommended for use in the classification of yeasts on the basis of their colonial characteristics and cell morphology.
  • Final pH (at 25 °C) is 5.6 ± 0.2
  • Cream to yellow homogeneous free flowing powder
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty