Would you like to log in again?
Yeast Carbon Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
M141-100G M141-100G
61000-024EA 96.46 USD InStock
CA61000-024 61000-024
Yeast Carbon Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Ideal for the classification of yeasts on the basis of their ability to assimilate nitrogen compounds.
  • Composed of a defined set of nutrients
  • Necessary vitamins included
  • Classification of yeasts
Yeast Carbon Base is composed of a defined set of nutrients including carbon source, amino acids, vitamins and minerals required for the growth of yeasts. The inclusion of vitamins in this base was found necessary as an aid forutilization of nitrogen compounds by certain yeasts as they cannot assimilate these compounds in the absence of vitamins.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty