Would you like to log in again?
Corn Meal Agar, HiMedia
Supplier: HiMedia Laboratories
M146-2.5KG M146-500G M146-500G M146-2.5KG M146-5KG
61000-028EA 494.83 USD InStock
CA61000-028 61000-028 CA89224-090 89224-090 89224-104
Corn Meal Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Corn meal agar is recommended for chlamydospore production by C. albicans and the maintenance of fungal stock cultures. Chlamydospore production is an accepted criterion for the identification of Candida species.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty