Would you like to log in again?
Yeast Nitrogen Base without Amino Acids and Ammonium Sulphate, Dehydrated, HiMedia Laboratories
Supplier: HiMedia Laboratories
M151-100G M151-100G
61000-034EA 122.51 USD InStock
CA61000-034 61000-034
Yeast Nitrogen Base without Amino Acids and Ammonium Sulphate, Dehydrated, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the classification of yeasts based on the ability to assimilate nitrogen and carbon compounds.
  • Microbiological culture media
  • 100 g poly bottle
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty