Would you like to log in again?
AATCC Mineral Salts Agar, HiMedia
Supplier: HiMedia Laboratories
M232-500G M232-500G
61000-056EA 299.36 USD InStock
CA61000-056 61000-056
AATCC Mineral Salts Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For evaluation of fungicides for use on textiles and resistance of textiles to mildew and rot.
Used as per the procedure described by American Association of Textile Chemists and Colourists for testing Initial mildew resistance of textiles, perseverance of mildew resistance, Fungicidal potency of textile fungicides testable, Standard Minimum protective concentration, peccnanence indices, including resistance to leaching, volatilization and weathering.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty