Would you like to log in again?
Algae Culture Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M342-500G M342-5KG M342-500G M342-2.5KG M342-2.5KG M342-5KG
61000-076EA 298.73 USD InStock
61000-076 89225-230 CA89225-228 89225-228 CA89225-230 CA61000-076
Algae Culture Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Algae Culture Broth is recommended for the isolation and cultivation of algae from soil, water and sewage.
  • White to light yellow homogeneous free flowing powder
  • White colored clear to slightly opalescent solution in tubes
  • Reaction of 0.187% w/v aqueous solution at 25 °C at pH 7.0±0.2
The medium provides all necessary nutrients for good growth of Algae but does not provide for other than minimal growth of bacteria and fungi.
Algae culture broth is also used to prepare the inoculum for the bioassay of algicidal chemicals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty