Would you like to log in again?
Bushnell Haas Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
M350-500G
61000-082EA 379.49 USD InStock
61000-082
Bushnell Haas Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Bushnell Hass broth is used for examining fuels for microbial contamination and for studying hydrocarbon deterioration by microorganisms.
  • Microbiological examination of fuels by the SIM Committee on microbiological deteriorations of fuels
  • Contain all nutrients except carbon source
  • Study hydrocarbon deterioration by microorganisms
Bushnell Haas Broth is prepared as per the formula of Bushnell and Haas and recommended for the microbiological examination of fuels by the SIM Committee on microbiological deteriorations of fuels
These bacteria can decompose a variety of hydrocarbons like kerosene, mineral oil, paraffin wax and gasoline. For liquid hydrocarbon the hydrocarbon is layered on the surface of inoculated agar. For testing volatile hydrocarbons such as gasoline the Petri-plates containing the medium are inverted and the hydrocarbon is poured into the lid. Magnesium sulphate, calcium chloride and ferric chloride provide trace elements. Ammonium nitrate is a nitrogen source while monopotassium phosphate and potassium phosphate buffers the medium.
These media contain all nutrients except carbon source, necessary for the growth of bacteria. Only those bacteria that are able to decompose hydrocarbon will grow in these media. Specific carbon source i.e. hydrocarbon can be added to this medium and their utilization by different microorganisms can be studied.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty