Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ISP Medium No. 7, Tyrosine Agar, HiMedia Laboratories

Supplier: HiMedia
M362-500G
61000-090EA 208.46 USD InStock
61000-090
ISP Medium No. 7, Tyrosine Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
ISP Medium No

  • Appearance: Cream to yellow homogeneous free flowing powder
  • Gelling: Gelling Firm, comparable with 2.0% agar gel
  • Colour and Clarity of prepared medium: Yellow coloured, clear to slightly opalescent gel forms in petri plates
  • pH
  • : 7.20 – 7.40
  • Cultural response: Cultural characteristics observed after an incubation at 25 to 30 °C for 48 – 72 hours

It is used for the differentiation of Sreptomyces species based on tyrosine utilization. The medium contains L-tyrosine, which is utilized by Streptomyces species.

7 (Tyrosine Agar) is recommended for the isolation and characterization of Streptomyces species.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty