Would you like to log in again?
Carbon Utilisation Agar, HiMedia
Supplier: HiMedia Laboratories
M363-500G M363-500G
61000-092EA 272.74 USD InStock
CA61000-092 61000-092
Carbon Utilisation Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
These are used for characterization of Streptomyces on the basis of carbon utilisation source.
Streptomyces are a group of gram-positive bacteria belonging to Actinobacteria found in soil and decaying vegetation. Carbon Utilization Agar is developed as per International Streptomyces Project for the cultivation and differentiation of Streptomyces purpureus and other Streptomyces species based on carbohydrate utilization. The various salts provide the electrolytes and minerals essential for the growth of Streptomyces species. The carbohydrates used for the studies are glucose, sucrose, xylose, arabinose, inositol, mannitol, fructose, rhamnose, raffinose or cellulose.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty