Would you like to log in again?
Folic Acid Casei Medium, HiMedia
Supplier: HiMedia Laboratories
M543-100G M543-100G
61000-114EA 375.87 USD InStock
CA61000-114 61000-114
Folic Acid Casei Medium, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Folic Acid Casei Medium is used for the microbiological assay of folic acid in blood serum using Lactobacillus casei ATCC 7469 as the test organism.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty