Would you like to log in again?
MY 40 Agar, Osmophilic Agar, Himedia
Supplier: HiMedia Laboratories
M594-500G M594-5KG M594-2.5KG M594-5KG M594-500G M594-2.5KG
61000-120EA 232.27 USD InStock
CA89225-958 61000-120 89225-956 CA89225-956 89225-958 CA61000-120
MY 40 Agar, Osmophilic Agar, Himedia
Media for Microbiology Dehydrated Media for Microbiology
Osmophilic Agar (MY 40 Agar) is generally used for detection of small numbers of osmotolerant yeasts in high-sugar foods and is useful for enumeration.
  • Contains malt and yeast extracts
  • 40% sucrose
  • Useful for enumeration
MY-40 Agar stands for malt extract and yeast extract and 40 for the 40% of sucrose in the medium.
The medium contains malt and yeast extracts, which supply the nitrogenous nutrients, amino acids, vitamins, trace ingredients to the osmophilic yeasts. 40% sucrose in the medium satisfy the nutritional need of the yeasts Aspergillus brasiliensis, Mucor racemosus, Pencillium notatum, Saccharomyces rouxii and results in good growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty