Would you like to log in again?
Toluidine Blue DNA Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M613-100G M613-100G M613-500G M613-500G
95022-074EA 1033.34 USD InStock
CA95022-074 CA61000-126 95022-074 61000-126
Toluidine Blue DNA Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Toluidine blue DNA agar is used for detection of thermostable deoxyribonuclease activity.
DNA in the medium enables the detection of DNase activity which depolymerizes the DNA resulting in the formation of a clear zone around the microbial growth. Inclusion of toluidine blue aids in detection of DNase activity by the production of a visible bright rose-pink coloured reaction due to its metachromatic properties. Tris amino methane forms the buffering system. Sodium chloride and calcium chloride provides the ions and also maintains osmotic equilibrium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty