Would you like to log in again?
Dehydrated Isolation Medium for Iron Bacteria, HiMedia Laboratories
Supplier: HiMedia Laboratories
M622-500G M622-500G M622-5KG M622-2.5KG M622-2.5KG M622-5KG
61000-130EA 204.42 USD InStock
61000-130 89226-054 CA89226-052 89226-052 CA89226-054 CA61000-130
Dehydrated Isolation Medium for Iron Bacteria, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Isolation medium for iron bacteria is used for the isolation of iron bacteria, especially those belonging to Sphaerotilus-Leptothrix group.
  • Store below 30 °C in tightly closed container and prepared medium at 2 to 8 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty