Would you like to log in again?
Mineral Modified Glutamate Medium Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
GM643-500G M643-500G M643-500G
61000-136EA 217.92 USD InStock
CA61000-136 61000-136 75804-366
Mineral Modified Glutamate Medium Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Base medium is used for enumeration of coliform bacteria in water and waste water.
  • Used for enumeration of coliforms in water and wastewater samples
  • Can also be used for detecting Escherichia coli in chlorinated waters
  • Contains a variety of nutrients including salts, amino acids and vitamins
Mineral Modified Glutamate Medium is used for enumeration of coliforms in water and wastewater samples. Folpmers first described a chemically defined medium based on glutamic acid for enumerating coliforms in water. However in the early days following its discovery, it was seen that the medium containing glucose gave many false positive results in 48 hours.It was then modified by Gray who incorporated formate and lactose in the medium which gave improved performance. This medium can also be used for detecting Escherichia coli in chlorinated waters and is better than Lauryl Tryptose Lactose Broth for the detection of small numbers of Escherichia coli. This medium contains a variety of nutrients including salts, amino acids and vitamins. Lactose is the fermentable carbohydrate and bromo cresol purple is the pH indicator. Because of the nutrients, this medium is superior for enumerating coliforms in water and wastewater as it satisfies most of the nutritional requirements of coliforms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty