Would you like to log in again?
Ashby’s Mannitol Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M706-2.5KG M706-5KG M706-500G M706-5KG M706-2.5KG M706-500G
89226-212EA 734.85 USD InStock
89226-212 CA89226-214 CA61000-154 CA89226-212 61000-154 89226-214
Ashby’s Mannitol Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Ashby's mannitol agar is used for cultivation of Azotobacter species that can use mannitol and atmospheric nitrogen as source of carbon and nitrogen respectively.
  • White to cream homogeneous free flowing powder
  • Firm, comparable with 1.5% Agar gel
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty