Would you like to log in again?
Limabean Agar, Himedia
Supplier: HiMedia Laboratories
M736-500G
61000-180EA 303.1 USD InStock
61000-180
Limabean Agar, Himedia
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation of phytopathological and other fungi.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty