Would you like to log in again?
Veg 8 Juice Broth, Himedia®
Supplier: HiMedia Laboratories
M895-500G M895-500G M895-2.5KG M895-5KG M895-2.5KG M895-5KG
89226-714EA 1005.57 USD InStock
89226-714 CA61000-208 61000-208 CA89226-716 CA89226-714 89226-716
Veg 8 Juice Broth, Himedia®
Media for Microbiology Dehydrated Media for Microbiology
Veg 8 juice broth is used for cultivation of fungi-yeasts and moulds.
  • Off-white to light yellow colored
  • Homogeneous, free flowing powder
  • Reaction of 2.43% w/v aqueous solution at 25 °C
Caution: Store dehydrated and the prepared medium at 2 to 8 °C in tightly closed container.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty