Would you like to log in again?
HiVeg™ Tryptone Glucose Extract Agar, HiMedia Laboratories
Supplier: HiMedia
MV014-5KG MV014-2.5KG MV014-2.5KG MV014-500G MV014-5KG MV014-500G
89227-196EA 626.12 USD InStock
89227-196 CA89227-196 CA61000-246 89227-198 CA89227-198 61000-246
HiVeg™ Tryptone Glucose Extract Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used for the enumeration of bacteria in water, air, milk and dairy products.
  • Light yellow colored
  • Homogeneous, free flowing powder
  • Reaction of 2.4% w/v aqueous solution at 25 °C
Caution: Store below 30 °C and the prepared medium at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty