Would you like to log in again?
Lauryl Sulphate HiVeg™ Broth, HiMedia
Supplier: HiMedia Laboratories
MV080-5KG MV080-5KG MV080-500G MV080-2.5KG MV080-2.5KG MV080-500G
61000-350EA 125.27 USD InStock
61000-350 89227-390 CA89227-390 89227-388 CA61000-350 CA89227-388
Lauryl Sulphate HiVeg™ Broth, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
These lauryl sulphate broth is for detection and enumeration of coliform bacteria in water, wastewater, dairy products and other food samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty