Would you like to log in again?
Lysine Arginine Iron HiVeg™ Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV1230-100G MV1230-100G
61000-570EA 113.47 USD InStock
61000-570 CA61000-570
Lysine Arginine Iron HiVeg™ Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Lysine Arginine Iron HiVeg™ Agar is used for the isolation and presumptive identification of Yersinia species from milk and milk products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty